Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 1 | 156 | ||
NM_001308340.1 | NP_001295269.1 | 1 | 156 | |||
GenBank mRNA | AK307955.1 | - | 1 | 111 | ||
AL712776.1 | - | 1 | 149 | |||
AY845858.1 | - | 1 | 17 | |||
BC137413.1 | - | 1 | 72 | 9021411 | ||
BC137414.1 | - | 1 | 72 | 9021412 | ||
BC144617.1 | - | 1 | 72 |