Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013620.3 | NP_001013642.1 | 3 | 890 | ||
XM_005268665.4 | XP_005268722.1 | 3 | 671 | |||
GenBank mRNA | AK307955.1 | - | 3 | 845 | ||
AY845858.1 | - | 3 | 751 | |||
BC137413.1 | - | 3 | 806 | 9021411 | ||
BC137414.1 | - | 3 | 806 | 9021412 | ||
BC144617.1 | - | 3 | 780 |