Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010857.2 | NP_001010857.1 | 2 | 255 | ||
GenBank mRNA | BC087854.1 | - | 2 | 195 | 30387074 | |
BC130507.1 | - | 2 | 235 | 40146538 | ||
BC130511.1 | - | 2 | 235 | 40146542 | ||
DB069578.1 | - | 2 | 255 | |||
DB086762.1 | - | 2 | 255 |