Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012337.2 | NP_001012337.1 | 6 | 1033 | ||
NM_001308313.1 | NP_001295242.1 | 7 | 1079 | |||
GenBank mRNA | AF231410.1 | - | 6 | 992 | ||
BC015413.1 | - | 4 | 773 | 4394326 | ||
BC141849.1 | - | 7 | 1026 | 8860347 |