Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019845.2 | NP_062819.1 | 1 | 506 | ||
GenBank mRNA | AB043585.1 | - | 1 | 506 | ||
AK074808.1 | - | 1 | 458 | |||
AK311958.1 | - | 1 | 366 | |||
BC002908.2 | - | 1 | 450 | 3942270 |