Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009811.3 | NP_001009811.2 | 19, 20 | 2682 | ||
NM_001319228.1 | NP_001306157.1 | 20, 21 | 2799 | |||
NM_001319229.1 | NP_001306158.1 | 19, 20 | 2691 | |||
NM_014984.3 | NP_055799.2 | 20, 21 | 2790 | |||
XM_011524547.2 | XP_011522849.1 | 20, 21 | 2596 | |||
XM_011524548.2 | XP_011522850.1 | 20, 21 | 2790 | |||
XM_011524550.2 | XP_011522852.1 | 20, 21 | 2799 | |||
XM_011524551.2 | XP_011522853.1 | 20, 21 | 2799 | |||
XM_011524552.2 | XP_011522854.1 | 19, 20 | 2691 | |||
XM_011524553.2 | XP_011522855.1 | 19, 20 | 2691 | |||
XM_011524555.2 | XP_011522857.1 | 20, 21 | 2799 | |||
XM_017024369.1 | XP_016879858.1 | 20, 21 | 2799 | |||
GenBank mRNA | AB029041.1 | - | 20, 21 | 2809 | ||
BC011615.2 | - | 11, 12 | 1531 | |||
BC136578.1 | - | 20, 21 | 2731 | 9020568 | ||
BC136580.1 | - | 20, 21 | 2731 | 9020570 | ||
BE732160.1 | - | 2, 3 | 205 | |||
BE734213.1 | - | 2, 3 | 207 | |||
BG773915.1 | - | 2, 3 | 227 |