Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024843.1 | NP_001020014.1 | 24 | 6199 | ||
NM_001162501.1 | NP_001155973.1 | 23 | 8520 | |||
NM_015088.2 | NP_055903.2 | 21 | 8190 |