Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012753.1 | NP_001012771.1 | 4 | 2392 | ||
GenBank mRNA | AK092240.1 | - | 4 | 2392 |
Gene Symbol: ZNF69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027231.1 | XP_016882720.1 | 5 | 762 |