Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013734.2 | NP_001013756.2 | 3 | 692 | ||
XM_017010897.1 | XP_016866386.1 | 4 | 834 | |||
XM_017010898.1 | XP_016866387.1 | 4 | 1009 | |||
GenBank mRNA | AK122906.1 | - | 3 | 692 | ||
BC132796.1 | - | 2 | 523 | 40146818 | ||
BC132798.1 | - | 2 | 523 | 40146820 |