Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017437.3 | NP_001017437.2 | 6 | 1727 | ||
NM_001318334.1 | NP_001305263.1 | 6 | 1773 | |||
XM_006724278.3 | XP_006724341.1 | 6 | 1334 | |||
XM_006724279.3 | XP_006724342.1 | 6 | 1334 | |||
XM_006724280.3 | XP_006724343.1 | 6 | 1334 | |||
XM_011530263.2 | XP_011528565.1 | 6 | 1387 | |||
XM_011530266.2 | XP_011528568.1 | 6 | 1334 | |||
XM_011530267.2 | XP_011528569.1 | 6 | 1334 | |||
XM_011530268.2 | XP_011528570.1 | 6 | 1334 | |||
XM_011530269.2 | XP_011528571.1 | 2 | 104 | |||
XM_011530270.2 | XP_011528572.1 | 6 | 1334 | |||
GenBank mRNA | BC092419.1 | - | 6 | 1361 | 30383661 | |
CR936833.1 | - | 4 | 1688 |