Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VANGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020335.2 | NP_065068.1 | 8 | 2794 | ||
XM_005245357.1 | XP_005245414.1 | 8 | 2601 | |||
XM_011509804.1 | XP_011508106.1 | 8 | 2649 | |||
GenBank mRNA | AB033041.1 | - | 7 | 2470 |