Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF618 SIRNA Assays ›
Gene Symbol:
ZNF618
Gene Name:
zinc finger protein 618
Gene Aliases:
FP13169, NEDD10
Chromosome Location:
Chr.9: 113876282 - 114056595 on Build GRCh38
UniGene ID:
Hs.349208
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
279817
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF618
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001318040.1
NP_001304969.1
13, 14
1357
NM_001318041.1
NP_001304970.1
13, 14
1360
NM_001318042.1
NP_001304971.1
14, 15
1456
NM_133374.2
NP_588615.2
13, 14
1177
XM_005251692.2
XP_005251749.1
16, 17
1558
XM_005251699.2
XP_005251756.1
13, 14
1210
XM_011518200.1
XP_011516502.1
15, 16
1522
XM_011518201.1
XP_011516503.1
15, 16
1498
XM_011518203.2
XP_011516505.1
15, 16
1450
XM_011518204.2
XP_011516506.1
14, 15
1390
XM_011518209.2
XP_011516511.1
3, 4
314
XM_017014241.1
XP_016869730.1
16, 17
1555
XM_017014242.1
XP_016869731.1
15, 16
1513
XM_017014243.1
XP_016869732.1
15, 16
1510
XM_017014244.1
XP_016869733.1
14, 15
1450
XM_017014245.1
XP_016869734.1
14, 15
1405
XM_017014246.1
XP_016869735.1
14, 15
1402
XM_017014247.1
XP_016869736.1
15, 16
8161
XM_017014248.1
XP_016869737.1
15, 16
1318
XM_017014249.1
XP_016869738.1
15, 16
1315
XM_017014250.1
XP_016869739.1
14, 15
1270
XM_017014251.1
XP_016869740.1
14, 15
9547
XM_017014252.1
XP_016869741.1
13, 14
1213
GenBank mRNA
AB075832.1
-
5, 6
1370
BC036039.1
-
15, 16
1459
5270564
BC053892.1
-
13, 14
1110
BC136287.1
-
13, 14
1305
9020274
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-sq4fc:80/100.66.76.194:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline