Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029858.3 | NP_001025029.2 | 8 | 1814 | ||
GenBank mRNA | AK128717.1 | - | 6 | 1162 | ||
BC028615.1 | - | 8 | 1817 | 4817165 | ||
BC036493.1 | - | 8 | 1582 | |||
BC114925.1 | - | 9 | 1688 | 40025808 |