Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRHDE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026837.1 | 1 | 1324 | |||
GenBank mRNA | AK095156.1 | - | 1 | 341 | ||
BC093901.1 | - | 1 | 250 | |||
BC093903.1 | - | 1 | 250 | |||
DA139014.1 | - | 1 | 380 |