Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253723.1 | NP_001240652.1 | 2 | 230 | ||
NM_001253724.1 | NP_001240653.1 | 2 | 233 | |||
NM_020134.3 | NP_064519.2 | 2 | 313 | |||
GenBank mRNA | AF157634.1 | - | 1 | 155 | ||
AF264015.1 | - | 2 | 317 | |||
AJ251275.1 | - | 1 | 155 | |||
BC002874.2 | - | 2 | 230 | 3940393 | ||
BT006871.1 | - | 1 | 155 | |||
DA410946.1 | - | 2 | 233 |