Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025357.2 | NP_001020528.1 | 1, 2 | 348 | ||
GenBank mRNA | AB181519.1 | - | 1, 2 | 329 | ||
BC140924.1 | - | 1, 2 | 374 | 9056736 | ||
BC142621.1 | - | 1, 2 | 348 | 40147448 |