Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020137.4 | NP_064522.3 | 14, 15 | 1219 | ||
XM_011543935.1 | XP_011542237.1 | 14, 15 | 1219 | |||
XM_011543936.1 | XP_011542238.1 | 14, 15 | 1219 | |||
XM_011543937.1 | XP_011542239.1 | 14, 15 | 1266 | |||
XM_017029656.1 | XP_016885145.1 | 14, 15 | 1219 | |||
XM_017029657.1 | XP_016885146.1 | 14, 15 | 1590 | |||
XM_017029658.1 | XP_016885147.1 | 14, 15 | 2529 | |||
XM_017029659.1 | XP_016885148.1 | 14, 15 | 1266 | |||
XM_017029660.1 | XP_016885149.1 | 14, 15 | 1620 | |||
XM_017029661.1 | XP_016885150.1 | 14, 15 | 1266 | |||
GenBank mRNA | AB032993.1 | - | 14, 15 | 1164 | ||
AK292158.1 | - | 14, 15 | 1190 | |||
AL136847.1 | - | 1, 2 | 105 | |||
BC001522.2 | - | 4, 5 | 332 | |||
BC101544.1 | - | 14, 15 | 1217 | 8069050 | ||
BC101546.1 | - | 14, 15 | 1217 | 8069052 | ||
BX640704.1 | - | 8, 9 | 903 | |||
BX647801.1 | - | 13, 14 | 1059 | |||
BX647804.1 | - | 13, 14 | 1059 |