Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168368.1 | NP_001161840.1 | 15 | 3623 | ||
NM_020692.2 | NP_065743.2 | 16 | 2462 | |||
XM_017021498.1 | XP_016876987.1 | 17 | 2449 | |||
GenBank mRNA | AK091426.1 | - | 1 | 1756 | ||
BC036812.1 | - | 16 | 2213 | 5734009 | ||
BC098578.1 | - | 16 | 2228 | 30924466 |