Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3GLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287045.1 | NP_001273974.1 | 2 | 279 | ||
NM_001287046.1 | NP_001273975.1 | 2 | 279 | |||
NM_020145.3 | NP_064530.1 | 2 | 279 | |||
XM_005252098.2 | XP_005252155.1 | 2 | 247 | |||
XM_005252100.2 | XP_005252157.1 | 2 | 246 | |||
XM_006717188.2 | XP_006717251.1 | 2 | 251 | |||
XM_006717189.2 | XP_006717252.1 | 2 | 246 | |||
XM_006717190.1 | XP_006717253.1 | 2 | 251 | |||
XM_006717191.1 | XP_006717254.1 | 2 | 251 | |||
GenBank mRNA | AB058751.2 | - | 2 | 205 | ||
AF257319.1 | - | 2 | 140 | |||
AF258589.1 | - | 2 | 192 | |||
BC014635.1 | - | 2 | 226 | 3677306 | ||
BM808895.1 | - | 290 |