Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030059.2 | NP_001025230.1 | 7 | 1093 | ||
NM_001318166.1 | NP_001305095.1 | 6 | 968 | |||
NM_001318167.1 | NP_001305096.1 | 5 | 904 | |||
NM_001318168.1 | NP_001305097.1 | 4 | 813 | |||
NM_001318169.1 | NP_001305098.1 | 3 | 642 | |||
NR_134516.1 | 5 | 605 | ||||
XM_005269592.2 | XP_005269649.1 | 6 | 922 | |||
XM_006717685.3 | XP_006717748.1 | 7 | 762 | |||
XM_011539444.2 | XP_011537746.1 | 8 | 820 | |||
XM_017015821.1 | XP_016871310.1 | 4 | 503 | |||
XM_017015822.1 | XP_016871311.1 | 4 | 470 | |||
GenBank mRNA | AK098668.1 | - | 7 | 676 | ||
BC101267.2 | - | 5 | 611 | |||
BC101268.2 | - | 6 | 598 | 40024099 | ||
BC101269.1 | - | 5 | 488 | |||
BC122535.1 | - | 4 | 439 | |||
BC132787.1 | - | 7 | 916 | 40146809 | ||
DB473853.2 | - | 6 | 675 |