Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SV2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297716.1 | NP_001284645.1 | 6, 7 | 1336 | ||
NM_014979.3 | NP_055794.3 | 6, 7 | 1336 | |||
XM_011543281.2 | XP_011541583.1 | 6, 7 | 1703 | |||
XM_011543282.2 | XP_011541584.2 | 6, 7 | 2263 | |||
GenBank mRNA | AB028977.1 | - | 4, 5 | 397 | ||
AK094917.1 | - | 6, 7 | 1337 | |||
BC100824.1 | - | 6, 7 | 1272 | 40001296 | ||
BC100825.1 | - | 6, 7 | 1270 | 40001297 | ||
BC100826.1 | - | 6, 7 | 1270 | 40001302 | ||
BC100827.1 | - | 6, 7 | 1270 | 40001303 |