Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102725072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135922.1 | 6 | 3688 | |||
GenBank mRNA | BC051721.1 | - | 6 | 3688 |
Gene Symbol: POM121L8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024583.1 | 7 | 4749 | |||
GenBank mRNA | AL117485.1 | - | 5 | 2700 | ||
BC050331.1 | - | 2 | 933 | |||
BC068032.1 | - | 2 | 794 | |||
BC090873.1 | - | 2 | 654 |