Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037671.3 | NP_001032760.1 | 3 | 1462 | ||
NM_001178097.2 | NP_001171568.1 | 2 | 1973 | |||
GenBank mRNA | BC043363.1 | - | 2 | 1788 | ||
BC043364.1 | - | 2 | 1788 | |||
BC105728.1 | - | 3 | 1242 | 40025557 | ||
BC107813.1 | - | 3 | 1241 | 40025555 |