Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034842.3 | NP_001030014.2 | 1, 2 | 1215 | ||
GenBank mRNA | AK126025.1 | - | 1, 2 | 1215 | ||
BC107139.2 | - | 1, 2 | 1139 | 40026539 | ||
JF332167.1 | - | 1, 2 | 1097 |