Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020872.2 | NP_065923.1 | 22 | 4338 | ||
XM_005264757.3 | XP_005264814.1 | 23 | 4448 | |||
XM_011533768.2 | XP_011532070.1 | 22 | 7970 | |||
XM_017006507.1 | XP_016861996.1 | 23 | 5018 | |||
XM_017006508.1 | XP_016861997.1 | 22 | 4243 | |||
XM_017006509.1 | XP_016861998.1 | 13 | 3017 | |||
GenBank mRNA | AB040929.1 | - | 20 | 3987 | ||
AK001157.1 | - | 1 | 318 | |||
AL512746.1 | - | 2 | 1363 |