Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135920.2 | NP_001129392.2 | 9 | 1118 | ||
NM_001206557.1 | NP_001193486.1 | 9 | 1118 | |||
NM_020829.3 | NP_065880.2 | 9 | 1118 | |||
XM_005251523.3 | XP_005251580.1 | 9 | 1204 | |||
XM_011517967.2 | XP_011516269.1 | 2 | 116 | |||
XM_017014934.1 | XP_016870423.1 | 9 | 774 | |||
XM_017014935.1 | XP_016870424.1 | 7 | 1675 | |||
XM_017014936.1 | XP_016870425.1 | 7 | 614 | |||
GenBank mRNA | AB205401.1 | - | 8 | 712 | ||
AK124394.1 | - | 6 | 1646 | |||
AL136875.1 | - | 9 | 910 | |||
BC136616.1 | - | 8 | 712 | 9020606 | ||
BC144296.1 | - | 8 | 712 | 9052816 |