Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135920.2 | NP_001129392.2 | 15, 16 | 1879 | ||
NM_001206557.1 | NP_001193486.1 | 14, 15 | 1768 | |||
NM_020829.3 | NP_065880.2 | 15, 16 | 1879 | |||
XM_005251523.3 | XP_005251580.1 | 15, 16 | 1965 | |||
XM_011517967.2 | XP_011516269.1 | 8, 9 | 877 | |||
XM_011517968.2 | XP_011516270.1 | 3, 4 | 212 | |||
XM_017014934.1 | XP_016870423.1 | 15, 16 | 1535 | |||
XM_017014935.1 | XP_016870424.1 | 13, 14 | 2436 | |||
XM_017014936.1 | XP_016870425.1 | 13, 14 | 1375 | |||
GenBank mRNA | AB205401.1 | - | 14, 15 | 1473 | ||
AK022642.1 | - | 1, 2 | 99 | |||
AK124394.1 | - | 11, 12 | 2387 | |||
AL136875.1 | - | 15, 16 | 1671 | |||
BC136616.1 | - | 14, 15 | 1473 | 9020606 | ||
BC144296.1 | - | 13, 14 | 1362 | 9052816 |