Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308133.1 | NP_001295062.1 | 9 | 1720 | ||
NM_001308134.1 | NP_001295063.1 | 10 | 1798 | |||
NM_020937.3 | NP_065988.1 | 10 | 1798 | |||
XM_011537034.2 | XP_011535336.1 | 10 | 1797 | |||
XM_011537035.2 | XP_011535337.1 | 9 | 1719 | |||
XM_017021523.1 | XP_016877012.1 | 10 | 1797 | |||
XM_017021524.1 | XP_016877013.1 | 8 | 1019 | |||
XM_017021525.1 | XP_016877014.1 | 8 | 1586 | |||
XM_017021526.1 | XP_016877015.1 | 7 | 830 | |||
XM_017021527.1 | XP_016877016.1 | 9 | 1611 | |||
GenBank mRNA | AK093422.1 | - | 10 | 1797 | ||
BC036056.1 | - | 10 | 1799 | |||
BC140776.1 | - | 9 | 1705 | 9021644 | ||
BC144511.1 | - | 9 | 1705 | 9053038 | ||
DQ140356.1 | - | 10 | 1805 |