Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258031.1 | NP_001244960.1 | 5 | 723 | ||
NM_001258032.1 | NP_001244961.1 | 5 | 764 | |||
GenBank mRNA | AK097682.1 | - | 3 | 329 | ||
BC136669.1 | - | 3 | 560 | 9020659 | ||
BX647438.1 | - | 5 | 764 | |||
DQ103711.1 | - | 5 | 724 |