Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301860.1 | NP_001288789.1 | 18 | 2365 | ||
NR_126051.1 | 18 | 2283 | ||||
XM_011528996.1 | XP_011527298.1 | 18 | 2531 | |||
XM_011528997.2 | XP_011527299.1 | 19 | 2737 | |||
XM_011528998.2 | XP_011527300.1 | 18 | 2554 | |||
XM_011528999.2 | XP_011527301.1 | 19 | 2684 | |||
XM_011529000.1 | XP_011527302.1 | 18 | 2528 | |||
XM_011529001.1 | XP_011527303.1 | 17 | 2210 | |||
XM_011529002.2 | XP_011527304.1 | 17 | 2333 | |||
XM_011529003.1 | XP_011527305.1 | 18 | 2701 | |||
XM_011529004.1 | XP_011527306.1 | 19 | 2422 | |||
XM_017028020.1 | XP_016883509.1 | 18 | 2360 | |||
GenBank mRNA | BC140806.1 | - | 18 | 2246 | 9021674 | |
BX647082.1 | - | 15 | 3343 | |||
BX648609.1 | - | 1 | 98 |