Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037668.1 | NP_001032757.2 | 2 | 317 | ||
GenBank mRNA | BC137510.1 | - | 2 | 243 | 9021510 | |
BC137513.1 | - | 2 | 243 | 9021513 | ||
DQ119826.1 | - | 2 | 317 |
Gene Symbol: DEFB107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040705.1 | NP_001035795.1 | 2 | 317 |