Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037162.1 | NP_001032239.1 | 2 | 971 | ||
XM_011537076.2 | XP_011535378.1 | 3 | 1578 | |||
GenBank mRNA | DQ082756.1 | - | 2 | 971 |