Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102405.1 | 1, 2 | 339 | |||
GenBank mRNA | AK307809.1 | - | 1, 2 | 119 | ||
AY894567.1 | - | 1, 2 | 339 | |||
AY894568.1 | - | 2, 3 | 237 | |||
AY894569.1 | - | 2, 3 | 239 | |||
AY894571.1 | - | 1, 2 | 339 | |||
AY894573.1 | - | 1, 2 | 339 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110864.1 | 5, 6 | 1205 | |||
GenBank mRNA | AF419617.1 | - | 5, 6 | 708 | ||
BX648497.1 | - | 5, 6 | 1004 |