Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020184.3 | NP_064569.3 | 1 | 604 | ||
XM_005263914.3 | XP_005263971.1 | 1 | 521 | |||
XM_005263915.3 | XP_005263972.1 | 1 | 521 | |||
XM_011510955.2 | XP_011509257.1 | 1 | 521 | |||
XM_011510956.2 | XP_011509258.1 | 1 | 521 | |||
GenBank mRNA | AB046812.1 | - | 1 | 334 | ||
AF202777.1 | - | 1 | 365 | |||
BC063295.1 | - | 1 | 604 | 30340626 |