Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133547.4 | NP_598231.3 | 10 | 2371 | ||
XM_006244242.2 | XP_006244304.1 | 9 | 2349 | |||
XM_006244243.2 | XP_006244305.1 | 9 | 2341 | |||
XM_006244245.2 | XP_006244307.1 | 9 | 2336 | |||
XM_006244246.2 | XP_006244308.1 | 9 | 2324 | |||
XM_006244248.2 | XP_006244310.1 | 10 | 2466 | |||
XM_006244249.2 | XP_006244311.1 | 8 | 2275 | |||
XM_006244250.2 | XP_006244312.1 | 8 | 2384 | |||
XM_008766738.1 | XP_008764960.1 | 9 | 2354 | |||
XM_008766739.1 | XP_008764961.1 | 9 | 2317 | |||
XM_008766740.1 | XP_008764962.1 | 9 | 2347 | |||
XM_008766741.1 | XP_008764963.1 | 9 | 2342 | |||
XM_008766742.1 | XP_008764964.1 | 9 | 2329 | |||
XM_008766743.1 | XP_008764965.1 | 9 | 2372 | |||
XM_008766744.1 | XP_008764966.1 | 8 | 2262 | |||
XM_008766745.1 | XP_008764967.1 | 8 | 2250 | |||
GenBank mRNA | BC103636.1 | - | 10 | 2371 | 7106590 |
Gene Symbol: LOC100910526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244254.2 | XP_006244316.1 | 2184 |
Gene Symbol: RGD1562392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750614.3 | XP_003750662.1 | 8 | 2282 | ||
XM_008766791.1 | XP_008765013.1 | 8 | 2261 |