Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012696.2 | NP_036828.2 | 1242 | |||
GenBank mRNA | BC087028.1 | - | 1242 | 7130388 | ||
FQ211035.1 | - | 650 | ||||
FQ211199.1 | - | 648 | ||||
FQ219050.1 | - | 648 | ||||
FQ219322.1 | - | 1124 | ||||
FQ219531.1 | - | 650 | ||||
FQ219669.1 | - | 649 | ||||
M11883.1 | - | 1012 | ||||
X02299.1 | - | 977 |