Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300942.1 | NP_001287871.1 | 3 | 229 | ||
NM_001300943.1 | NP_001287872.1 | 3 | 229 | |||
NM_001300944.1 | NP_001287873.1 | 3 | 229 | |||
NM_020193.4 | NP_064578.2 | 3 | 229 | |||
XM_005274112.4 | XP_005274169.1 | 3 | 326 | |||
XM_006718637.3 | XP_006718700.1 | 3 | 326 | |||
XM_006718638.3 | XP_006718701.1 | 3 | 239 | |||
XM_006718639.3 | XP_006718702.1 | 3 | 239 | |||
XM_011545173.2 | XP_011543475.1 | 3 | 326 | |||
XM_011545174.2 | XP_011543476.1 | 3 | 326 | |||
XM_011545175.2 | XP_011543477.1 | 3 | 239 | |||
XM_011545177.2 | XP_011543479.1 | 3 | 326 | |||
XM_011545178.2 | XP_011543480.1 | 3 | 326 | |||
XM_011545179.2 | XP_011543481.1 | 3 | 239 | |||
XM_017018021.1 | XP_016873510.1 | 3 | 326 | |||
XM_017018022.1 | XP_016873511.1 | 2 | 126 | |||
GenBank mRNA | AJ430203.1 | - | 2 | 86 | ||
AK125114.1 | - | 3 | 195 | |||
AK304043.1 | - | 3 | 229 | |||
AK309621.1 | - | 3 | 223 | |||
BC033404.1 | - | 3 | 229 | |||
BC117265.1 | - | 2 | 125 | 40125816 | ||
BC143370.1 | - | 2 | 125 | 9051875 | ||
BC143374.1 | - | 2 | 125 | 9051879 | ||
BC143376.1 | - | 2 | 125 | 9051881 | ||
BG722092.1 | - | 3 | 234 |