Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000017.1 | NP_001000017.1 | 1 | 754 |
Gene Symbol: LOC100910876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767751.1 | XP_008765973.1 | 754 |