Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.535192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2804
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001402.5
NP_001393.1
3
351
XM_011535514.2
XP_011533816.1
3
986
GenBank mRNA
AF174496.1
-
2
181
AF328730.1
-
1
59
AF397403.1
-
1
96
AJ420488.1
-
2
310
AK024888.1
-
1
349
AK026650.1
-
1
351
AK055058.1
-
3
351
AK058051.1
-
3
351
AK058082.1
-
3
351
AK058163.1
-
2
169
AK098510.1
-
3
351
AK098518.1
-
3
211
AK098623.1
-
3
351
AK127384.1
-
3
351
AK222515.1
-
1
351
AK222523.1
-
1
351
AK222551.1
-
1
351
AK222982.1
-
1
351
AK223030.1
-
1
348
AK223042.1
-
1
206
AK223046.1
-
1
354
AK289374.1
-
3
351
AK289442.1
-
3
289
AK290691.1
-
3
351
AK292639.1
-
3
667
AK297878.1
-
3
350
AK300922.1
-
3
212
AK304214.1
-
3
352
AY043301.1
-
2
698
BC008587.1
-
1
337
3447654
BC009733.1
-
1
301
3877516
BC009875.2
-
1
341
3948601
BC010735.1
-
1
330
3898364
BC012891.1
-
1
329
3858483
BC014224.2
-
1
340
4335756
BC018150.1
-
1
337
3851939
BC018641.2
-
1
337
3660140
BC018850.2
-
1
340
BC019050.2
-
1
303
BC021686.1
-
1
356
3930863
BC028674.1
-
1
355
4107346
BC029337.1
-
1
325
BC029343.1
-
1
323
BC029997.1
-
1
337
BC035877.1
-
1
352
BC038339.1
-
1
337
3461588
BC057391.1
-
1
306
4339606
BC066893.1
-
1
351
4818863
BC071619.1
-
1
330
4448926
BC071727.1
-
1
306
4514479
BC071741.1
-
1
331
3885539
BC071841.1
-
1
326
4587981
BC072385.1
-
1
338
5585092
BC082268.1
-
1
330
6729595
BC111051.1
-
1
341
5585079
DQ867534.1
-
1
37
EF219140.1
-
3
511
EF362804.1
-
3
581
X03558.1
-
3
341
X16869.1
-
2
303
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbzcs:80/10.239.166.68:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline