Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019617.3 | NP_062563.3 | 4, 5 | 419 | ||
GenBank mRNA | AB039886.1 | - | 4, 5 | 419 | ||
AJ966789.1 | - | 4, 5 | 419 | |||
AY139184.1 | - | 4, 5 | 357 | |||
AY359050.1 | - | 4, 5 | 343 | |||
AY456961.1 | - | 4, 5 | 356 | |||
BC059778.1 | - | 4, 5 | 345 |