Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008931.1 | NP_001008931.1 | 39 | |||
GenBank mRNA | AY510315.1 | - | 39 |
Gene Symbol: LOC103691006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758859.1 | XP_008757081.1 | 1 | 56 |