Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008972.1 | NP_001008972.1 | 1 | 428 | ||
GenBank mRNA | AY510283.1 | - | 1 | 428 |
Gene Symbol: LOC100909473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236570.1 | XP_006236632.1 | 1 | 428 |