Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly49si1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009497.1 | NP_001009497.1 | 3 | 345 | ||
XM_008763312.1 | XP_008761534.1 | 3 | 596 | |||
XM_008763313.1 | XP_008761535.1 | 4 | 626 | |||
XM_008763314.1 | XP_008761536.1 | 4 | 420 | |||
XM_008763315.1 | XP_008761537.1 | 3 | 596 | |||
XM_008763316.1 | XP_008761538.1 | 3 | 596 | |||
XM_008763317.1 | XP_008761539.1 | 2 | 506 | |||
XM_008763318.1 | XP_008761540.1 | 2 | 506 | |||
XM_008763319.1 | XP_008761541.1 | 3 | 596 | |||
GenBank mRNA | AY659935.1 | - | 3 | 345 |