Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009628.1 | NP_001009628.1 | 9, 10 | 1210 | ||
GenBank mRNA | BC088161.1 | - | 9, 10 | 1210 | 7369906 | |
FQ211311.1 | - | 7, 8 | 773 | |||
FQ219207.1 | - | 7, 8 | 772 | |||
FQ219641.1 | - | 7, 8 | 774 | |||
M11661.1 | - | 9, 10 | 1108 | |||
M11885.1 | - | 4, 5 | 408 |