Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144002.1 | NP_001137474.1 | 4 | 1142 | ||
NM_019857.4 | NP_062831.3 | 4 | 695 | |||
NM_175859.2 | NP_787055.1 | 4 | 781 | |||
XM_005274562.3 | XP_005274619.1 | 4 | 558 | |||
XM_006724503.3 | XP_006724566.1 | 5 | 733 | |||
XM_011545545.2 | XP_011543847.1 | 4 | 554 | |||
GenBank mRNA | AF226667.1 | - | 4 | 598 | ||
AK023549.1 | - | 4 | 653 | |||
AK024070.1 | - | 4 | 636 | |||
AK125332.1 | - | 4 | 1142 | |||
AK125348.1 | - | 4 | 772 | |||
BC006256.2 | - | 2 | 137 | |||
BC034986.1 | - | 4 | 760 | 5268973 |