Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013053.1 | NP_001013071.1 | 1 | 137 | ||
GenBank mRNA | AF115768.1 | - | 1 | 98 |