Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012739.1 | NP_001012757.1 | 509 | |||
NM_001160231.1 | NP_001153703.1 | 504 | ||||
NM_001160232.1 | NP_001153704.1 | 506 | ||||
GenBank mRNA | BC091402.1 | - | 509 | 7324328 | ||
FQ211511.1 | - | 499 | ||||
FQ214844.1 | - | 490 | ||||
FQ221164.1 | - | 494 | ||||
FQ223418.1 | - | 499 | ||||
FQ226183.1 | - | 546 | ||||
FQ231235.1 | - | 499 | ||||
X62671.1 | - | 499 |
Gene Symbol: LOC100360647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750172.3 | XP_003750220.1 | 1 | 485 |
Gene Symbol: LOC686658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771420.1 | XP_008769642.1 | 1 | 530 |
Gene Symbol: LOC687780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750169.3 | XP_003750217.1 | 1 | 485 |