Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318502.1 | NP_001305431.1 | 1 | 1044 | ||
NM_019892.5 | NP_063945.2 | 1 | 1044 | |||
XM_017014926.1 | XP_016870415.1 | 1 | 1083 | |||
GenBank mRNA | AF187891.1 | - | 1 | 745 | ||
BC028032.1 | - | 1 | 1026 | 5242186 | ||
BC110356.1 | - | 1 | 1045 | |||
BE314630.1 | - | 1 | 228 |