Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024990.2 | NP_001020161.2 | 2 | 197 | ||
XM_008767565.1 | XP_008765787.1 | 2 | 288 | |||
GenBank mRNA | BC091278.1 | - | 2 | 187 | ||
CO388545.1 | - | 2 | 197 |