Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024303.1 | NP_001019474.1 | 7 | 1162 | ||
XM_006232978.2 | XP_006233040.1 | 6 | 1182 | |||
GenBank mRNA | BC087077.1 | - | 7 | 1162 | 7135157 |